Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003910.3 | NP_003901.2 | 4 - 5 | 647 | 126 |
XM_005250670.4 | XP_005250727.1 | 4 - 5 | 669 | 126 | |
XM_005250671.4 | XP_005250728.1 | 4 - 5 | 743 | 126 | |
XM_005250674.3 | XP_005250731.1 | 4 - 5 | 764 | 126 | |
XM_017012760.1 | XP_016868249.1 | 4 - 5 | 479 | 126 | |
XM_017012761.1 | XP_016868250.1 | 4 - 5 | 642 | 126 | |
XM_017012762.1 | XP_016868251.1 | 4 - 5 | 479 | 126 | |
GenBank mRNA | AK297784.1 | - | 4 - 5 | 415 | 126 |
AK316477.1 | - | 4 - 5 | 646 | 126 | |
BC022821.1 | - | 4 - 5 | 636 | 126 | |
BC104670.1 | - | 4 - 5 | 419 | 126 | |
CR456951.1 | - | 2 - 3 | 214 | 126 |