Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007349.3 | NP_031375.3 | 7 - 8 | 1844 | 92 |
XM_005249539.1 | XP_005249596.1 | 8 - 9 | 1983 | 92 | |
XM_011515982.2 | XP_011514284.1 | 7 - 8 | 1958 | 92 | |
XM_011515983.1 | XP_011514285.1 | 7 - 8 | 1844 | 92 | |
XM_017011881.1 | XP_016867370.1 | 8 - 9 | 3156 | 92 | |
GenBank mRNA | AK093584.1 | - | 1 - 2 | 538 | 92 |
AK123044.1 | - | 3 - 4 | 1889 | 92 | |
AK123600.1 | - | 8 - 9 | 1983 | 92 | |
AK293744.1 | - | 5 - 6 | 1676 | 92 | |
AK307417.1 | - | 7 - 8 | 1844 | 92 | |
BC033781.1 | - | 2 - 3 | 1085 | 92 | |
BX538201.1 | - | 1 - 2 | 620 | 92 | |
BX640616.1 | - | 7 - 8 | 3742 | 92 | |
U80735.1 | - | 3 - 4 | 827 | 92 |