Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74457109-74457346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNE3
Entrez Gene ID
10008
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 3
Gene Aliases
HOKPP, HYPP, MiRP2
UniGene
Hs.523899
Gene Chromosome Location
Chr.11: 74454841-74467729 on Build GRCh38
Transcripts
NM_005472.4 XM_011544713.2 XM_017017047.1 XM_017017048.1 XM_017017049.1 XM_017017050.1 XM_017017051.1 XM_017017052.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6318635 c.295C>A p.R99S Chr.11: 74457269-74457269 on Build GRCh38
6690771 c.291G>T p.K97N Chr.11: 74457273-74457273 on Build GRCh38
4413997 c.259A>T p.K87* Chr.11: 74457305-74457305 on Build GRCh38
71254 c.263G>T p.R88L Chr.11: 74457301-74457301 on Build GRCh38
1746579 c.288C>T p.I96I Chr.11: 74457276-74457276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41312923 Chr.11: 74457194-74457194 on Build GRCh38
rs1055293593 Chr.11: 74457200-74457200 on Build GRCh38
rs199972628 Chr.11: 74457285-74457285 on Build GRCh38
rs771943960 Chr.11: 74457280-74457280 on Build GRCh38
rs755835033 Chr.11: 74457249-74457249 on Build GRCh38
rs955717859 Chr.11: 74457167-74457167 on Build GRCh38
rs993767466 Chr.11: 74457231-74457231 on Build GRCh38
rs764717543 Chr.11: 74457214-74457214 on Build GRCh38
rs888537773 Chr.11: 74457269-74457269 on Build GRCh38
rs923090414 Chr.11: 74457159-74457159 on Build GRCh38
rs141695803 Chr.11: 74457254-74457254 on Build GRCh38
rs867623173 Chr.11: 74457130-74457130 on Build GRCh38
rs746750241 Chr.11: 74457300-74457300 on Build GRCh38
rs11822977 Chr.11: 74457235-74457235 on Build GRCh38
rs768106696 Chr.11: 74457327-74457327 on Build GRCh38
rs1006962870 Chr.11: 74457290-74457290 on Build GRCh38
rs542842233 Chr.11: 74457323-74457323 on Build GRCh38
rs530360248 Chr.11: 74457201-74457201 on Build GRCh38
rs557750053 Chr.11: 74457234-74457234 on Build GRCh38
rs924962054 Chr.11: 74457180-74457180 on Build GRCh38
rs786205809 Chr.11: 74457284-74457284 on Build GRCh38
rs762915607 Chr.11: 74457206-74457206 on Build GRCh38
rs927526096 Chr.11: 74457221-74457221 on Build GRCh38
rs945325355 Chr.11: 74457169-74457169 on Build GRCh38
rs11236111 Chr.11: 74457172-74457172 on Build GRCh38
rs968424822 Chr.11: 74457197-74457197 on Build GRCh38
rs763157740 Chr.11: 74457302-74457302 on Build GRCh38
rs966495313 Chr.11: 74457317-74457317 on Build GRCh38
rs375856416 Chr.11: 74457303-74457303 on Build GRCh38
rs368585507 Chr.11: 74457248-74457248 on Build GRCh38
rs976125449 Chr.11: 74457308-74457308 on Build GRCh38
rs978419298 Chr.11: 74457322-74457322 on Build GRCh38
rs535453362 Chr.11: 74457205-74457205 on Build GRCh38
rs565798552 Chr.11: 74457204-74457204 on Build GRCh38
rs964790424 Chr.11: 74457306-74457306 on Build GRCh38
rs556647231 Chr.11: 74457260-74457260 on Build GRCh38
rs531245825 Chr.11: 74457129-74457129 on Build GRCh38
rs753448731 Chr.11: 74457257-74457257 on Build GRCh38
rs201425581 Chr.11: 74457202-74457202 on Build GRCh38
rs748088627 Chr.11: 74457269-74457269 on Build GRCh38
rs779555364 Chr.11: 74457251-74457251 on Build GRCh38
rs773675377 Chr.11: 74457230-74457230 on Build GRCh38
rs121908441 Chr.11: 74457268-74457268 on Build GRCh38
rs17221833 Chr.11: 74457301-74457301 on Build GRCh38
rs17215437 Chr.11: 74457316-74457316 on Build GRCh38

View Full Product Details