Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017820.4 | NP_060290.3 | 21 - 22 | 2511 | 159 |
XM_011518810.1 | XP_011517112.1 | 23 - 24 | 2976 | 159 | |
XM_011518814.1 | XP_011517116.1 | 15 - 16 | 1926 | 159 | |
XM_011518815.1 | XP_011517117.1 | 12 - 13 | 1610 | 159 | |
XM_017014861.1 | XP_016870350.1 | 24 - 25 | 3120 | 159 | |
XM_017014862.1 | XP_016870351.1 | 22 - 23 | 2861 | 159 | |
XM_017014863.1 | XP_016870352.1 | 22 - 23 | 2819 | 159 | |
XM_017014864.1 | XP_016870353.1 | 21 - 22 | 3181 | 159 | |
XM_017014865.1 | XP_016870354.1 | 21 - 22 | 2828 | 159 | |
XM_017014866.1 | XP_016870355.1 | 20 - 21 | 2484 | 159 | |
XM_017014867.1 | XP_016870356.1 | 20 - 21 | 2525 | 159 | |
GenBank mRNA | AK055004.1 | - | 11 - 12 | 1643 | 159 |
AK094438.1 | - | 21 - 22 | 2411 | 159 | |
BU947354.1 | - | 3 - 4 | 233 | 159 |