Assay Design



Gene Transcripts

Gene Symbol
EXD3
Entrez Gene ID
54932
Gene Name
exonuclease 3'-5' domain containing 3
Gene Aliases
mut-7
Chromosome Location
Chr.9: 137306894 - 137423262 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017820.4 NP_060290.3 10 - 11 1064 90
XM_017014863.1 XP_016870352.1 11 - 12 1279 90
GenBank mRNA AK094438.1 - 10 - 11 964 90

View Full Product Details