Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130896.2 | NP_570966.2 | 3 - 4 | 354 | 70 |
NM_181510.2 | NP_852611.2 | 3 - 4 | 354 | 70 | |
XM_017028119.1 | XP_016883608.1 | 3 - 4 | 333 | 70 | |
GenBank mRNA | AF492015.1 | - | 3 - 4 | 354 | 70 |
AF492016.1 | - | 3 - 4 | 354 | 70 | |
AL591715.1 | - | 2 - 3 | 249 | 70 | |
GQ472209.1 | - | 3 - 4 | 354 | 70 |