Assay Design



Gene Transcripts

Gene Symbol
KRT73
Entrez Gene ID
319101
Gene Name
keratin 73
Gene Aliases
IRT6IRS3, K6IRS3, KRT6IRS3
Chromosome Location
Chr.12: 52601813 - 52620133 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175068.2 NP_778238.1 8 - 9 1402 111
XM_011538257.2 XP_011536559.1 7 - 8 2880 111
GenBank mRNA AJ508776.1 - 8 - 9 1402 111
BC109212.2 - 7 - 8 1760 111

View Full Product Details