Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13563633-13563878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN2B
Entrez Gene ID
2904
Gene Name
glutamate ionotropic receptor NMDA type subunit 2B
Gene Aliases
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3
UniGene
Hs.504844
Gene Chromosome Location
Chr.12: 13537337-13982012 on Build GRCh38
Transcripts
NM_000834.3 XM_005253351.3 XM_011520628.2 XM_011520629.2 XM_017019219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6136274 c.3383G>T p.G1128V Chr.12: 13563855-13563855 on Build GRCh38
6519990 c.3423G>T p.E1141D Chr.12: 13563815-13563815 on Build GRCh38
2002688 c.3564C>T p.G1188G Chr.12: 13563674-13563674 on Build GRCh38
2002692 c.3466A>G p.K1156E Chr.12: 13563772-13563772 on Build GRCh38
1705187 c.3493G>A p.D1165N Chr.12: 13563745-13563745 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746001325 Chr.12: 13563743-13563743 on Build GRCh38
rs535509676 Chr.12: 13563673-13563673 on Build GRCh38
rs199852144 Chr.12: 13563667-13563667 on Build GRCh38
rs200439105 Chr.12: 13563856-13563856 on Build GRCh38
rs200237781 Chr.12: 13563823-13563823 on Build GRCh38
rs1017679931 Chr.12: 13563674-13563674 on Build GRCh38
rs750952904 Chr.12: 13563774-13563774 on Build GRCh38
rs201250088 Chr.12: 13563747-13563747 on Build GRCh38
rs267603392 Chr.12: 13563859-13563859 on Build GRCh38
rs748196005 Chr.12: 13563708-13563708 on Build GRCh38
rs764171423 Chr.12: 13563748-13563748 on Build GRCh38
rs1042339 Chr.12: 13563739-13563739 on Build GRCh38
rs764845072 Chr.12: 13563781-13563781 on Build GRCh38
rs762050994 Chr.12: 13563763-13563763 on Build GRCh38
rs774809461 Chr.12: 13563754-13563754 on Build GRCh38
rs1006226313 Chr.12: 13563672-13563672 on Build GRCh38
rs767870767 Chr.12: 13563702-13563702 on Build GRCh38
rs866581841 Chr.12: 13563741-13563741 on Build GRCh38
rs148625092 Chr.12: 13563849-13563849 on Build GRCh38
rs45600931 Chr.12: 13563740-13563740 on Build GRCh38
rs187979330 Chr.12: 13563825-13563825 on Build GRCh38
rs373548904 Chr.12: 13563776-13563776 on Build GRCh38
rs777639067 Chr.12: 13563733-13563733 on Build GRCh38
rs979079013 Chr.12: 13563838-13563838 on Build GRCh38
rs957733664 Chr.12: 13563658-13563658 on Build GRCh38
rs141886903 Chr.12: 13563686-13563686 on Build GRCh38
rs757924519 Chr.12: 13563724-13563724 on Build GRCh38
rs267603391 Chr.12: 13563664-13563664 on Build GRCh38
rs758094582 Chr.12: 13563805-13563805 on Build GRCh38
rs775478008 Chr.12: 13563746-13563746 on Build GRCh38
rs143955920 Chr.12: 13563765-13563765 on Build GRCh38
rs769658975 Chr.12: 13563855-13563855 on Build GRCh38
rs747800865 Chr.12: 13563716-13563716 on Build GRCh38
rs866476716 Chr.12: 13563695-13563695 on Build GRCh38
rs202120141 Chr.12: 13563752-13563752 on Build GRCh38
rs201861590 Chr.12: 13563661-13563661 on Build GRCh38
rs746865690 Chr.12: 13563830-13563830 on Build GRCh38
rs1806191 Chr.12: 13563704-13563704 on Build GRCh38
rs202195729 Chr.12: 13563736-13563736 on Build GRCh38
rs752200554 Chr.12: 13563782-13563782 on Build GRCh38
rs201568782 Chr.12: 13563689-13563689 on Build GRCh38
rs200166566 Chr.12: 13563692-13563692 on Build GRCh38
rs951406434 Chr.12: 13563725-13563725 on Build GRCh38
rs556194434 Chr.12: 13563710-13563710 on Build GRCh38
rs774794141 Chr.12: 13563714-13563714 on Build GRCh38
rs367543149 Chr.12: 13563676-13563676 on Build GRCh38
rs756986518 Chr.12: 13563831-13563831 on Build GRCh38
rs183502070 Chr.12: 13563734-13563734 on Build GRCh38
rs937786796 Chr.12: 13563853-13563853 on Build GRCh38
rs942708350 Chr.12: 13563742-13563742 on Build GRCh38
rs769857006 Chr.12: 13563745-13563745 on Build GRCh38

View Full Product Details