Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291661.1 | NP_001278590.1 | 60 - 61 | 9865 | 133 |
XM_011533466.1 | XP_011531768.1 | 60 - 61 | 9850 | 133 | |
XM_011533467.1 | XP_011531769.1 | 60 - 61 | 9817 | 133 | |
XM_011533468.1 | XP_011531770.1 | 60 - 61 | 9787 | 133 | |
XM_017005860.1 | XP_016861349.1 | 60 - 61 | 9850 | 133 | |
XM_017005861.1 | XP_016861350.1 | 59 - 60 | 10533 | 133 | |
XM_017005862.1 | XP_016861351.1 | 39 - 40 | 6425 | 133 | |
GenBank mRNA | AK097746.1 | - | 1 - 2 | 151 | 133 |
AK128592.1 | - | 8 - 9 | 1291 | 133 |