Assay Design
Gene Transcripts
- Gene Symbol
- STRA8
- Entrez Gene ID
-
346673
- Gene Name
- stimulated by retinoic acid 8
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 135231979 - 135258661 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_182489.1 |
NP_872295.1 |
1 - 2 |
62 |
150 |
GenBank mRNA |
AF513502.1 |
- |
1 - 2 |
62 |
150 |
View Full Product Details