Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182500.2 | NP_872306.1 | 2 - 3 | 579 | 65 |
XM_006711857.2 | XP_006711920.1 | 2 - 3 | 503 | 65 | |
XM_006711858.2 | XP_006711921.1 | 2 - 3 | 503 | 65 | |
XM_006711859.3 | XP_006711922.1 | 2 - 3 | 503 | 65 | |
GenBank mRNA | AK057872.1 | - | 2 - 3 | 585 | 65 |
AK292828.1 | - | 2 - 3 | 504 | 65 | |
BC120930.2 | - | 2 - 3 | 546 | 65 |