Assay Design



Gene Transcripts

Gene Symbol
YIPF7
Entrez Gene ID
285525
Gene Name
Yip1 domain family member 7
Gene Aliases
FinGER9
Chromosome Location
Chr.4: 44619578 - 44656868 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182592.2 NP_872398.2 2 - 3 209 138
GenBank mRNA AK096895.1 - 2 - 3 209 138
BC103996.1 - 2 - 3 209 138

View Full Product Details