Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134468.1 | NP_001127940.1 | 3 - 4 | 396 | 91 |
NM_182597.2 | NP_872403.1 | 3 - 4 | 602 | 91 | |
XM_011516074.1 | XP_011514376.1 | 3 - 4 | 1105 | 91 | |
XM_011516075.2 | XP_011514377.1 | 3 - 4 | 2792 | 91 | |
XM_011516076.2 | XP_011514378.1 | 3 - 4 | 2428 | 91 | |
XM_017012028.1 | XP_016867517.1 | 2 - 3 | 2310 | 91 | |
GenBank mRNA | AK096894.1 | - | 3 - 4 | 618 | 91 |
BC031976.1 | - | 5 - 6 | 669 | 92 | |
BC126149.1 | - | 3 - 4 | 440 | 91 | |
BC130568.1 | - | 3 - 4 | 440 | 91 | |
DB456503.2 | - | 3 - 4 | 396 | 91 |