Assay Design



Gene Transcripts

Gene Symbol
KRTAP13-3
Entrez Gene ID
337960
Gene Name
keratin associated protein 13-3
Gene Aliases
KAP13.3
Chromosome Location
Chr.21: 30425393 - 30425912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181622.1 NP_853653.1 1 - 1 342 106
GenBank mRNA AB096940.1 - 1 - 1 356 106
BC103965.2 - 1 - 1 342 106

View Full Product Details