Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005335.1 | NP_001005335.1 | 11 - 12 | 1399 | 86 |
NM_001533.2 | NP_001524.2 | 11 - 12 | 1633 | 86 | |
XM_011526886.1 | XP_011525188.1 | 11 - 12 | 1422 | 86 | |
XM_011526887.1 | XP_011525189.1 | 11 - 12 | 1466 | 86 | |
XM_011526888.2 | XP_011525190.1 | 9 - 10 | 1521 | 86 | |
XM_011526889.1 | XP_011525191.1 | 11 - 12 | 1517 | 86 | |
GenBank mRNA | AB044547.1 | - | 11 - 12 | 1628 | 86 |
AK092828.1 | - | 5 - 6 | 1932 | 86 | |
AK094966.1 | - | 4 - 5 | 2623 | 86 | |
AK097975.1 | - | 3 - 4 | 1038 | 86 | |
AK292115.1 | - | 11 - 12 | 1398 | 86 | |
AK313652.1 | - | 11 - 12 | 1568 | 86 | |
BC069184.1 | - | 11 - 12 | 1572 | 86 | |
X16135.1 | - | 11 - 12 | 1557 | 86 |