Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005335.1 | NP_001005335.1 | 12 - 13 | 1495 | 75 |
NM_001533.2 | NP_001524.2 | 12 - 13 | 1729 | 75 | |
XM_011526886.1 | XP_011525188.1 | 12 - 13 | 1518 | 75 | |
XM_011526887.1 | XP_011525189.1 | 12 - 13 | 1562 | 75 | |
XM_011526888.2 | XP_011525190.1 | 10 - 11 | 1617 | 75 | |
XM_011526889.1 | XP_011525191.1 | 12 - 13 | 1613 | 75 | |
GenBank mRNA | AB044547.1 | - | 12 - 13 | 1724 | 75 |
AK092828.1 | - | 6 - 7 | 2028 | 75 | |
AK094966.1 | - | 5 - 6 | 2719 | 75 | |
AK292115.1 | - | 12 - 13 | 1494 | 75 | |
AK313652.1 | - | 12 - 13 | 1664 | 75 | |
BC069184.1 | - | 12 - 13 | 1668 | 75 | |
X16135.1 | - | 12 - 13 | 1653 | 75 |