Assay Design
Gene Transcripts
- Gene Symbol
- ABT1
- Entrez Gene ID
-
29777
- Gene Name
- activator of basal transcription 1
- Gene Aliases
-
hABT1
- Chromosome Location
-
Chr.6: 26596943 - 26600050 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_013375.3 |
NP_037507.1 |
3 - 3 |
1758 |
96 |
View Full Product Details