Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58545679-58545948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIPC
Entrez Gene ID
3990
Gene Name
lipase C, hepatic type
Gene Aliases
HDLCQ12, HL, HTGL, LIPH
UniGene
Hs.654472
Gene Chromosome Location
Chr.15: 58410754-58568952 on Build GRCh38
Transcripts
NM_000236.2 XM_005254372.1 XM_005254374.4 XM_006720502.3 XM_017022176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4557338 c.724G>A p.D242N Chr.15: 58545891-58545891 on Build GRCh38
6350477 c.591A>G p.G197G Chr.15: 58545758-58545758 on Build GRCh38
5788844 c.594T>A p.P198P Chr.15: 58545761-58545761 on Build GRCh38
5021833 c.672C>G p.T224T Chr.15: 58545839-58545839 on Build GRCh38
4553914 c.605G>A p.G202E Chr.15: 58545772-58545772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767030496 Chr.15: 58545849-58545849 on Build GRCh38
rs749271888 Chr.15: 58545925-58545926 on Build GRCh38
rs575306493 Chr.15: 58545841-58545841 on Build GRCh38
rs200684324 Chr.15: 58545790-58545790 on Build GRCh38
rs756970343 Chr.15: 58545879-58545879 on Build GRCh38
rs6082 Chr.15: 58545758-58545758 on Build GRCh38
rs756693135 Chr.15: 58545898-58545898 on Build GRCh38
rs939351730 Chr.15: 58545860-58545860 on Build GRCh38
rs751167742 Chr.15: 58545718-58545718 on Build GRCh38
rs755192110 Chr.15: 58545885-58545885 on Build GRCh38
rs774643284 Chr.15: 58545910-58545910 on Build GRCh38
rs34351746 Chr.15: 58545755-58545755 on Build GRCh38
rs780399994 Chr.15: 58545804-58545804 on Build GRCh38
rs752531604 Chr.15: 58545714-58545714 on Build GRCh38
rs781040430 Chr.15: 58545753-58545753 on Build GRCh38
rs777385339 Chr.15: 58545721-58545721 on Build GRCh38
rs751119878 Chr.15: 58545759-58545759 on Build GRCh38
rs897052423 Chr.15: 58545728-58545728 on Build GRCh38
rs770215498 Chr.15: 58545753-58545753 on Build GRCh38
rs781489450 Chr.15: 58545886-58545886 on Build GRCh38
rs527610957 Chr.15: 58545922-58545922 on Build GRCh38
rs769598517 Chr.15: 58545863-58545863 on Build GRCh38
rs748164947 Chr.15: 58545893-58545893 on Build GRCh38
rs199772942 Chr.15: 58545819-58545819 on Build GRCh38
rs766977535 Chr.15: 58545778-58545778 on Build GRCh38
rs545537175 Chr.15: 58545840-58545840 on Build GRCh38
rs371573112 Chr.15: 58545906-58545906 on Build GRCh38
rs754648703 Chr.15: 58545737-58545738 on Build GRCh38
rs749318726 Chr.15: 58545730-58545730 on Build GRCh38
rs542765160 Chr.15: 58545789-58545789 on Build GRCh38
rs558189607 Chr.15: 58545712-58545712 on Build GRCh38
rs760729045 Chr.15: 58545861-58545861 on Build GRCh38
rs778111687 Chr.15: 58545905-58545905 on Build GRCh38
rs6084 Chr.15: 58545839-58545839 on Build GRCh38
rs759018685 Chr.15: 58545762-58545762 on Build GRCh38
rs777246707 Chr.15: 58545737-58545737 on Build GRCh38
rs749932377 Chr.15: 58545904-58545905 on Build GRCh38
rs752656256 Chr.15: 58545781-58545781 on Build GRCh38
rs765084049 Chr.15: 58545880-58545880 on Build GRCh38
rs781089971 Chr.15: 58545713-58545713 on Build GRCh38
rs759143056 Chr.15: 58545846-58545846 on Build GRCh38
rs374328131 Chr.15: 58545754-58545754 on Build GRCh38
rs6083 Chr.15: 58545811-58545811 on Build GRCh38
rs775120008 Chr.15: 58545923-58545923 on Build GRCh38
rs747067984 Chr.15: 58545731-58545731 on Build GRCh38
rs753712250 Chr.15: 58545876-58545876 on Build GRCh38
rs373470233 Chr.15: 58545726-58545726 on Build GRCh38
rs747125166 Chr.15: 58545808-58545808 on Build GRCh38
rs777800802 Chr.15: 58545923-58545923 on Build GRCh38
rs748695211 Chr.15: 58545825-58545825 on Build GRCh38
rs199615899 Chr.15: 58545785-58545785 on Build GRCh38
rs752584800 Chr.15: 58545754-58545755 on Build GRCh38
rs2414592 Chr.15: 58545711-58545711 on Build GRCh38
rs141216569 Chr.15: 58545727-58545727 on Build GRCh38
rs199787635 Chr.15: 58545882-58545882 on Build GRCh38
rs745521832 Chr.15: 58545916-58545916 on Build GRCh38
rs149322349 Chr.15: 58545752-58545752 on Build GRCh38
rs780165647 Chr.15: 58545719-58545719 on Build GRCh38
rs752078494 Chr.15: 58545854-58545854 on Build GRCh38
rs770200473 Chr.15: 58545833-58545833 on Build GRCh38
rs954993789 Chr.15: 58545745-58545745 on Build GRCh38
rs6080 Chr.15: 58545734-58545734 on Build GRCh38
rs777626514 Chr.15: 58545787-58545787 on Build GRCh38
rs773685048 Chr.15: 58545836-58545836 on Build GRCh38
rs1029349848 Chr.15: 58545739-58545739 on Build GRCh38
rs771312009 Chr.15: 58545907-58545907 on Build GRCh38

View Full Product Details