Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 51054590-51054851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD4
Entrez Gene ID
4089
Gene Name
SMAD family member 4
Gene Aliases
DPC4, JIP, MADH4, MYHRS
UniGene
Hs.75862
Gene Chromosome Location
Chr.18: 51030213-51085042 on Build GRCh38
Transcripts
NM_005359.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
14104 c.1_*del p.0 Chr.18: 51047047-51078467 on Build GRCh38
6784674 c.507_508insCCAT p.L172fs*5 Chr.18: 51054833-51054834 on Build GRCh38
1151777 c.514T>A p.L172M Chr.18: 51054840-51054840 on Build GRCh38
7148725 c.499G>C p.E167Q Chr.18: 51054825-51054825 on Build GRCh38
6896806 c.474G>A p.V158V Chr.18: 51054800-51054800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759341259 Chr.18: 51054759-51054759 on Build GRCh38
rs1015033235 Chr.18: 51054682-51054682 on Build GRCh38
rs961773590 Chr.18: 51054720-51054721 on Build GRCh38
rs760578129 Chr.18: 51054731-51054731 on Build GRCh38
rs786201939 Chr.18: 51054795-51054797 on Build GRCh38
rs786201120 Chr.18: 51054809-51054809 on Build GRCh38
rs569300819 Chr.18: 51054628-51054628 on Build GRCh38
rs780716382 Chr.18: 51054797-51054797 on Build GRCh38
rs751763157 Chr.18: 51054783-51054783 on Build GRCh38
rs188806041 Chr.18: 51054639-51054639 on Build GRCh38
rs749594930 Chr.18: 51054800-51054800 on Build GRCh38
rs750573971 Chr.18: 51054744-51054745 on Build GRCh38
rs864622209 Chr.18: 51054786-51054786 on Build GRCh38
rs786203155 Chr.18: 51054810-51054810 on Build GRCh38
rs1014723442 Chr.18: 51054693-51054693 on Build GRCh38
rs199790852 Chr.18: 51054790-51054790 on Build GRCh38
rs1057519259 Chr.18: 51054789-51054789 on Build GRCh38
rs748927421 Chr.18: 51054748-51054748 on Build GRCh38
rs975802248 Chr.18: 51054721-51054721 on Build GRCh38
rs760712048 Chr.18: 51054658-51054658 on Build GRCh38
rs534355764 Chr.18: 51054792-51054792 on Build GRCh38
rs778555013 Chr.18: 51054827-51054828 on Build GRCh38
rs876660058 Chr.18: 51054817-51054817 on Build GRCh38
rs756675590 Chr.18: 51054796-51054796 on Build GRCh38
rs768452351 Chr.18: 51054753-51054753 on Build GRCh38
rs753617681 Chr.18: 51054755-51054755 on Build GRCh38
rs149541014 Chr.18: 51054738-51054738 on Build GRCh38
rs181178864 Chr.18: 51054775-51054775 on Build GRCh38

View Full Product Details