Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 87245545-87245802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP2B
Entrez Gene ID
25978
Gene Name
charged multivesicular body protein 2B
Gene Aliases
ALS17, CHMP2.5, DMT1, VPS2-2, VPS2B
UniGene
Hs.476930
Gene Chromosome Location
Chr.3: 87227263-87255548 on Build GRCh38
Transcripts
NM_001244644.1 NM_014043.3 XM_011533576.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7422859 c.143A>C p.K48T Chr.3: 87245730-87245730 on Build GRCh38
82067 c.192A>G p.Q64Q Chr.3: 87245779-87245779 on Build GRCh38
1425577 c.194T>G p.L65R Chr.3: 87245781-87245781 on Build GRCh38
335197 c.205C>T p.R69W Chr.3: 87245792-87245792 on Build GRCh38
2828050 c.151A>G p.K51E Chr.3: 87245738-87245738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011933799 Chr.3: 87245591-87245591 on Build GRCh38
rs201132171 Chr.3: 87245707-87245707 on Build GRCh38
rs376170805 Chr.3: 87245666-87245666 on Build GRCh38
rs566585457 Chr.3: 87245582-87245582 on Build GRCh38
rs143800051 Chr.3: 87245763-87245763 on Build GRCh38
rs953962504 Chr.3: 87245628-87245628 on Build GRCh38
rs767024180 Chr.3: 87245703-87245703 on Build GRCh38
rs775173904 Chr.3: 87245671-87245671 on Build GRCh38
rs769672519 Chr.3: 87245669-87245669 on Build GRCh38
rs757039889 Chr.3: 87245587-87245587 on Build GRCh38
rs894786074 Chr.3: 87245696-87245696 on Build GRCh38
rs745754634 Chr.3: 87245774-87245774 on Build GRCh38
rs188471297 Chr.3: 87245750-87245750 on Build GRCh38
rs548180396 Chr.3: 87245576-87245576 on Build GRCh38
rs989337134 Chr.3: 87245630-87245630 on Build GRCh38
rs915104269 Chr.3: 87245631-87245631 on Build GRCh38
rs778990578 Chr.3: 87245748-87245748 on Build GRCh38
rs534066538 Chr.3: 87245583-87245583 on Build GRCh38
rs761297424 Chr.3: 87245698-87245698 on Build GRCh38
rs755409238 Chr.3: 87245744-87245744 on Build GRCh38
rs762501606 Chr.3: 87245676-87245676 on Build GRCh38
rs538072644 Chr.3: 87245727-87245727 on Build GRCh38
rs369639551 Chr.3: 87245678-87245678 on Build GRCh38
rs374095325 Chr.3: 87245608-87245608 on Build GRCh38
rs762740572 Chr.3: 87245680-87245680 on Build GRCh38
rs974739906 Chr.3: 87245595-87245595 on Build GRCh38
rs777310161 Chr.3: 87245687-87245687 on Build GRCh38
rs575034954 Chr.3: 87245776-87245776 on Build GRCh38
rs774117372 Chr.3: 87245685-87245685 on Build GRCh38
rs148750997 Chr.3: 87245779-87245779 on Build GRCh38
rs756644379 Chr.3: 87245720-87245720 on Build GRCh38
rs758486323 Chr.3: 87245762-87245762 on Build GRCh38
rs376207785 Chr.3: 87245596-87245596 on Build GRCh38
rs373046970 Chr.3: 87245665-87245665 on Build GRCh38
rs754127865 Chr.3: 87245731-87245731 on Build GRCh38
rs370382342 Chr.3: 87245745-87245745 on Build GRCh38

View Full Product Details