Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74457293-74457538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNE3
Entrez Gene ID
10008
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 3
Gene Aliases
HOKPP, HYPP, MiRP2
UniGene
Hs.523899
Gene Chromosome Location
Chr.11: 74454841-74467729 on Build GRCh38
Transcripts
NM_005472.4 XM_011544713.2 XM_017017047.1 XM_017017048.1 XM_017017049.1 XM_017017050.1 XM_017017051.1 XM_017017052.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3935624 c.184C>G p.L62V Chr.11: 74457380-74457380 on Build GRCh38
4036674 c.167A>T p.N56I Chr.11: 74457397-74457397 on Build GRCh38
6838041 c.97C>T p.P33S Chr.11: 74457467-74457467 on Build GRCh38
1318090 c.83A>G p.N28S Chr.11: 74457481-74457481 on Build GRCh38
397958 c.126G>A p.Q42Q Chr.11: 74457438-74457438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530975076 Chr.11: 74457427-74457427 on Build GRCh38
rs745645715 Chr.11: 74457469-74457469 on Build GRCh38
rs780882536 Chr.11: 74457460-74457460 on Build GRCh38
rs780971201 Chr.11: 74457397-74457397 on Build GRCh38
rs1009916288 Chr.11: 74457385-74457385 on Build GRCh38
rs542842233 Chr.11: 74457323-74457323 on Build GRCh38
rs773287275 Chr.11: 74457515-74457515 on Build GRCh38
rs748308590 Chr.11: 74457472-74457472 on Build GRCh38
rs772286478 Chr.11: 74457481-74457481 on Build GRCh38
rs369134829 Chr.11: 74457391-74457391 on Build GRCh38
rs565287436 Chr.11: 74457406-74457406 on Build GRCh38
rs918843154 Chr.11: 74457512-74457512 on Build GRCh38
rs759641427 Chr.11: 74457497-74457497 on Build GRCh38
rs759214664 Chr.11: 74457428-74457428 on Build GRCh38
rs868057422 Chr.11: 74457419-74457419 on Build GRCh38
rs757890580 Chr.11: 74457442-74457442 on Build GRCh38
rs772767562 Chr.11: 74457414-74457414 on Build GRCh38
rs545135414 Chr.11: 74457396-74457396 on Build GRCh38
rs754598853 Chr.11: 74457368-74457368 on Build GRCh38
rs752159282 Chr.11: 74457373-74457373 on Build GRCh38
rs2270676 Chr.11: 74457366-74457366 on Build GRCh38
rs767752832 Chr.11: 74457518-74457518 on Build GRCh38
rs766125492 Chr.11: 74457358-74457358 on Build GRCh38
rs763497447 Chr.11: 74457449-74457450 on Build GRCh38
rs768106696 Chr.11: 74457327-74457327 on Build GRCh38
rs17221826 Chr.11: 74457336-74457336 on Build GRCh38
rs758367217 Chr.11: 74457382-74457382 on Build GRCh38
rs929457354 Chr.11: 74457491-74457491 on Build GRCh38
rs757172223 Chr.11: 74457451-74457451 on Build GRCh38
rs755300564 Chr.11: 74457470-74457470 on Build GRCh38
rs547172459 Chr.11: 74457495-74457495 on Build GRCh38
rs202036216 Chr.11: 74457349-74457349 on Build GRCh38
rs141880803 Chr.11: 74457444-74457444 on Build GRCh38
rs778413540 Chr.11: 74457369-74457369 on Build GRCh38
rs966495313 Chr.11: 74457317-74457317 on Build GRCh38
rs777664556 Chr.11: 74457383-74457383 on Build GRCh38
rs752253389 Chr.11: 74457438-74457438 on Build GRCh38
rs930193305 Chr.11: 74457516-74457516 on Build GRCh38
rs768960571 Chr.11: 74457402-74457402 on Build GRCh38
rs978419298 Chr.11: 74457322-74457322 on Build GRCh38
rs866853654 Chr.11: 74457415-74457415 on Build GRCh38
rs761179696 Chr.11: 74457424-74457424 on Build GRCh38
rs17215444 Chr.11: 74457425-74457425 on Build GRCh38
rs773590777 Chr.11: 74457421-74457421 on Build GRCh38
rs760938276 Chr.11: 74457342-74457342 on Build GRCh38
rs749496477 Chr.11: 74457399-74457399 on Build GRCh38
rs34604640 Chr.11: 74457448-74457448 on Build GRCh38
rs764834315 Chr.11: 74457430-74457430 on Build GRCh38
rs371666083 Chr.11: 74457407-74457407 on Build GRCh38
rs17215437 Chr.11: 74457316-74457316 on Build GRCh38

View Full Product Details