Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41610756-41610985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT16
Entrez Gene ID
3868
Gene Name
keratin 16
Gene Aliases
CK16, FNEPPK, K16, K1CP, KRT16A, NEPPK, PC1
UniGene
Hs.655160
Gene Chromosome Location
Chr.17: 41609778-41612827 on Build GRCh38
Transcripts
NM_005557.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416674 c.1042C>T p.Q348* Chr.17: 41610871-41610871 on Build GRCh38
4979021 c.969C>T p.S323S Chr.17: 41610944-41610944 on Build GRCh38
6146498 c.963C>A p.S321S Chr.17: 41610950-41610950 on Build GRCh38
6996610 c.996G>T p.E332D Chr.17: 41610917-41610917 on Build GRCh38
6453005 c.988C>T p.R330C Chr.17: 41610925-41610925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755302919 Chr.17: 41610892-41610892 on Build GRCh38
rs201947780 Chr.17: 41610821-41610821 on Build GRCh38
rs201459436 Chr.17: 41610936-41610936 on Build GRCh38
rs140407898 Chr.17: 41610924-41610924 on Build GRCh38
rs762787273 Chr.17: 41610922-41610922 on Build GRCh38
rs189042832 Chr.17: 41610886-41610886 on Build GRCh38
rs1000136710 Chr.17: 41610860-41610860 on Build GRCh38
rs756227425 Chr.17: 41610859-41610859 on Build GRCh38
rs267604877 Chr.17: 41610867-41610867 on Build GRCh38
rs767786872 Chr.17: 41610841-41610841 on Build GRCh38
rs765819730 Chr.17: 41610957-41610957 on Build GRCh38
rs758099620 Chr.17: 41610900-41610900 on Build GRCh38
rs566540770 Chr.17: 41610911-41610911 on Build GRCh38
rs370949280 Chr.17: 41610895-41610895 on Build GRCh38
rs759945966 Chr.17: 41610944-41610944 on Build GRCh38
rs141910921 Chr.17: 41610903-41610903 on Build GRCh38
rs762021081 Chr.17: 41610840-41610840 on Build GRCh38
rs777606459 Chr.17: 41610902-41610902 on Build GRCh38
rs774568431 Chr.17: 41610836-41610836 on Build GRCh38
rs1022825792 Chr.17: 41610786-41610786 on Build GRCh38
rs550935583 Chr.17: 41610823-41610823 on Build GRCh38
rs941583326 Chr.17: 41610950-41610950 on Build GRCh38
rs770867452 Chr.17: 41610808-41610808 on Build GRCh38
rs201864040 Chr.17: 41610802-41610802 on Build GRCh38
rs766374250 Chr.17: 41610925-41610925 on Build GRCh38
rs376392652 Chr.17: 41610843-41610843 on Build GRCh38
rs747010520 Chr.17: 41610884-41610884 on Build GRCh38
rs748580087 Chr.17: 41610898-41610898 on Build GRCh38
rs143438105 Chr.17: 41610904-41610904 on Build GRCh38
rs776461226 Chr.17: 41610809-41610809 on Build GRCh38
rs748813482 Chr.17: 41610836-41610837 on Build GRCh38
rs775254361 Chr.17: 41610920-41610920 on Build GRCh38
rs147159853 Chr.17: 41610905-41610905 on Build GRCh38
rs754014788 Chr.17: 41610943-41610943 on Build GRCh38
rs766869930 Chr.17: 41610874-41610874 on Build GRCh38
rs769839631 Chr.17: 41610912-41610912 on Build GRCh38

View Full Product Details