Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 65967682-65967955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUSB
Entrez Gene ID
2990
Gene Name
glucuronidase beta
Gene Aliases
BG, MPS7
UniGene
Hs.255230
Gene Chromosome Location
Chr.7: 65960684-65982314 on Build GRCh38
Transcripts
NM_000181.3 NM_001284290.1 NM_001293104.1 NM_001293105.1 NR_120531.1 XM_005250297.3 XM_011516113.1 XM_011516114.1 XM_017012091.1 XM_017012092.1 XM_017012093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6265275 c.1586A>G p.Y529C Chr.7: 65967798-65967798 on Build GRCh38
6337082 c.1653+7A>G p.? Chr.7: 65967724-65967724 on Build GRCh38
1209225 c.1534G>A p.G512R Chr.7: 65967850-65967850 on Build GRCh38
3882052 c.1646T>C p.F549S Chr.7: 65967738-65967738 on Build GRCh38
6236796 c.1481C>T p.P494L Chr.7: 65967903-65967903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548880426 Chr.7: 65967902-65967902 on Build GRCh38
rs768292111 Chr.7: 65967731-65967731 on Build GRCh38
rs777838133 Chr.7: 65967925-65967925 on Build GRCh38
rs778095106 Chr.7: 65967713-65967713 on Build GRCh38
rs756256226 Chr.7: 65967760-65967761 on Build GRCh38
rs767826344 Chr.7: 65967831-65967831 on Build GRCh38
rs368053364 Chr.7: 65967764-65967764 on Build GRCh38
rs981387734 Chr.7: 65967854-65967854 on Build GRCh38
rs121918178 Chr.7: 65967900-65967900 on Build GRCh38
rs758492902 Chr.7: 65967791-65967793 on Build GRCh38
rs201361157 Chr.7: 65967931-65967931 on Build GRCh38
rs778161730 Chr.7: 65967857-65967858 on Build GRCh38
rs527753075 Chr.7: 65967930-65967930 on Build GRCh38
rs377519272 Chr.7: 65967767-65967767 on Build GRCh38
rs779693654 Chr.7: 65967722-65967722 on Build GRCh38
rs768343264 Chr.7: 65967873-65967873 on Build GRCh38
rs749695052 Chr.7: 65967714-65967714 on Build GRCh38
rs762859080 Chr.7: 65967779-65967779 on Build GRCh38
rs752845837 Chr.7: 65967851-65967851 on Build GRCh38
rs761204529 Chr.7: 65967749-65967749 on Build GRCh38
rs746198220 Chr.7: 65967869-65967869 on Build GRCh38
rs763054536 Chr.7: 65967920-65967920 on Build GRCh38
rs929372840 Chr.7: 65967721-65967721 on Build GRCh38
rs759880257 Chr.7: 65967823-65967823 on Build GRCh38
rs751397640 Chr.7: 65967802-65967802 on Build GRCh38
rs786205674 Chr.7: 65967798-65967798 on Build GRCh38
rs771219944 Chr.7: 65967715-65967715 on Build GRCh38
rs766339888 Chr.7: 65967797-65967797 on Build GRCh38
rs760449944 Chr.7: 65967887-65967887 on Build GRCh38
rs794726973 Chr.7: 65967850-65967850 on Build GRCh38
rs772768221 Chr.7: 65967917-65967917 on Build GRCh38
rs776242697 Chr.7: 65967740-65967740 on Build GRCh38
rs747719992 Chr.7: 65967903-65967903 on Build GRCh38
rs143495981 Chr.7: 65967904-65967904 on Build GRCh38
rs199694121 Chr.7: 65967852-65967852 on Build GRCh38
rs121918179 Chr.7: 65967863-65967863 on Build GRCh38
rs746721071 Chr.7: 65967730-65967730 on Build GRCh38
rs778349139 Chr.7: 65967856-65967856 on Build GRCh38
rs201928248 Chr.7: 65967857-65967857 on Build GRCh38
rs756694911 Chr.7: 65967703-65967703 on Build GRCh38
rs767730315 Chr.7: 65967799-65967799 on Build GRCh38
rs769788461 Chr.7: 65967761-65967761 on Build GRCh38

View Full Product Details