Product Details

Assay Reference Genome
Location

Chr.2:27367471 on build GRCh38
Cytoband
2p23.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8890
Gene Symbol
EIF2B4
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
Location
Chr.2:27364352-27370457 on build GRCh38
Assay Gene Location
Overlaps Intron 9 - Exon 9
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
EIF2B4 NM_001034116.1 NP_001029288.1
NM_001318965.1 NP_001305894.1
NM_001318966.1 NP_001305895.1
NM_001318967.1 NP_001305896.1
NM_001318968.1 NP_001305897.1
NM_001318969.1 NP_001305898.1
NM_015636.3 NP_056451.3
NM_172195.3 NP_751945.2
XM_006712132.1 XP_006712195.1
XM_011533147.2 XP_011531449.1
AB209532.1 3 2377 BAD92769.1
AF057699.1 AAQ13438.1
AF112207.1 AAF17195.1
AF218009.1 AAG17251.1
AJ011305.1 CAB57260.1
AJ011306.1 CAB57261.1
AK300734.1 9 1061
AL050109.1 CAB43277.1
BC001870.1 AAH01870.1
BC091502.1 AAH91502.1
JQ991007.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv833614 Chr2:27263559 - 27452213 on Build GRCh38 Gain SLC30A3 KRTCAP3 SNX17 PPM1G IFT172 MPV17 NRBP1 TRIM54 GTF3C2-AS1 FTH1P3 DNAJC5G LOC105374363 UCN EIF2B4 GTF3C2 ZNF513
esv3575209 Chr2:27334578 - 27389381 on Build GRCh38 Gain LOC105374363 SNX17 PPM1G EIF2B4 GTF3C2 GTF3C2-AS1 ZNF513
nsv581244 Chr2:27335385 - 27480628 on Build GRCh38 Loss LOC105374363 KRTCAP3 SNX17 PPM1G IFT172 NRBP1 EIF2B4 GTF3C2 GTF3C2-AS1 ZNF513 FTH1P3

View Full Product Details