Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41747136-41747409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHOX2B
Entrez Gene ID
8929
Gene Name
paired like homeobox 2b
Gene Aliases
NBLST2, NBPhox, PMX2B
UniGene
Hs.87202
Gene Chromosome Location
Chr.4: 41744082-41748970 on Build GRCh38
Transcripts
NM_003924.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1220473 c.407A>G p.D136G Chr.4: 41747371-41747371 on Build GRCh38
5743023 c.421C>T p.R141* Chr.4: 41747357-41747357 on Build GRCh38
4714163 c.395C>A p.A132D Chr.4: 41747383-41747383 on Build GRCh38
188141 c.406G>A p.D136N Chr.4: 41747372-41747372 on Build GRCh38
6946981 c.395C>T p.A132V Chr.4: 41747383-41747383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750372598 Chr.4: 41747335-41747335 on Build GRCh38
rs28939716 Chr.4: 41747357-41747357 on Build GRCh38
rs796072391 Chr.4: 41747228-41747228 on Build GRCh38
rs1023527620 Chr.4: 41747165-41747165 on Build GRCh38
rs749906655 Chr.4: 41747336-41747336 on Build GRCh38
rs749630768 Chr.4: 41747298-41747298 on Build GRCh38
rs560766845 Chr.4: 41747309-41747309 on Build GRCh38
rs200756467 Chr.4: 41747318-41747318 on Build GRCh38
rs749727192 Chr.4: 41747367-41747367 on Build GRCh38
rs779269711 Chr.4: 41747385-41747385 on Build GRCh38
rs536966714 Chr.4: 41747230-41747230 on Build GRCh38
rs751450012 Chr.4: 41747325-41747325 on Build GRCh38
rs375957944 Chr.4: 41747311-41747311 on Build GRCh38
rs114158743 Chr.4: 41747269-41747269 on Build GRCh38
rs748614674 Chr.4: 41747387-41747387 on Build GRCh38
rs772048230 Chr.4: 41747310-41747310 on Build GRCh38
rs897313850 Chr.4: 41747282-41747282 on Build GRCh38
rs28647582 Chr.4: 41747248-41747248 on Build GRCh38
rs779822865 Chr.4: 41747341-41747341 on Build GRCh38
rs147497096 Chr.4: 41747373-41747373 on Build GRCh38
rs764154532 Chr.4: 41747324-41747324 on Build GRCh38
rs766544036 Chr.4: 41747313-41747313 on Build GRCh38
rs538746212 Chr.4: 41747257-41747257 on Build GRCh38
rs914600624 Chr.4: 41747226-41747226 on Build GRCh38
rs764944729 Chr.4: 41747330-41747330 on Build GRCh38
rs923793307 Chr.4: 41747193-41747193 on Build GRCh38
rs1041649186 Chr.4: 41747274-41747274 on Build GRCh38
rs541256864 Chr.4: 41747295-41747295 on Build GRCh38
rs575087990 Chr.4: 41747204-41747204 on Build GRCh38
rs1003452907 Chr.4: 41747339-41747339 on Build GRCh38
rs766950465 Chr.4: 41747335-41747335 on Build GRCh38
rs1048148839 Chr.4: 41747287-41747287 on Build GRCh38
rs527856290 Chr.4: 41747196-41747196 on Build GRCh38
rs1042144543 Chr.4: 41747364-41747364 on Build GRCh38
rs543317710 Chr.4: 41747340-41747340 on Build GRCh38
rs956351167 Chr.4: 41747197-41747197 on Build GRCh38
rs567926130 Chr.4: 41747229-41747229 on Build GRCh38
rs148316114 Chr.4: 41747167-41747167 on Build GRCh38
rs757155646 Chr.4: 41747326-41747326 on Build GRCh38
rs769328215 Chr.4: 41747303-41747303 on Build GRCh38
rs572123616 Chr.4: 41747284-41747284 on Build GRCh38
rs761622789 Chr.4: 41747302-41747302 on Build GRCh38
rs370922121 Chr.4: 41747315-41747315 on Build GRCh38
rs762544075 Chr.4: 41747202-41747202 on Build GRCh38
rs1011041347 Chr.4: 41747320-41747320 on Build GRCh38
rs576512130 Chr.4: 41747254-41747254 on Build GRCh38
rs984230223 Chr.4: 41747245-41747245 on Build GRCh38
rs774705849 Chr.4: 41747307-41747307 on Build GRCh38
rs974569832 Chr.4: 41747223-41747223 on Build GRCh38
rs775194855 Chr.4: 41747183-41747183 on Build GRCh38

View Full Product Details