Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042679.1 | NP_001036144.1 | 2 - 3 | 298 | 80 |
NM_175744.4 | NP_786886.1 | 2 - 3 | 400 | 80 | |
GenBank mRNA | AK094474.1 | - | 2 - 3 | 1208 | 80 |
BC007245.1 | - | 2 - 3 | 95 | 80 | |
BC009177.2 | - | 2 - 3 | 95 | 80 | |
BQ231012.1 | - | 2 - 3 | 288 | 80 | |
CN289691.1 | - | 2 - 3 | 296 | 80 | |
CX872299.1 | - | 2 - 3 | 400 | 80 |