Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74917912-74918169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USH1G
Entrez Gene ID
124590
Gene Name
USH1 protein network component sans
Gene Aliases
ANKS4A, SANS
UniGene
Hs.376688
Gene Chromosome Location
Chr.17: 74916083-74923263 on Build GRCh38
Transcripts
NM_001282489.2 NM_173477.4 XM_011524296.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6588905 c.1383-1G>A p.? Chr.17: 74918077-74918077 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs113765180 Chr.17: 74918010-74918010 on Build GRCh38
rs779370429 Chr.17: 74918139-74918139 on Build GRCh38
rs146004909 Chr.17: 74918071-74918071 on Build GRCh38
rs746580796 Chr.17: 74918081-74918081 on Build GRCh38
rs748104851 Chr.17: 74918103-74918103 on Build GRCh38
rs757954601 Chr.17: 74918074-74918074 on Build GRCh38
rs764308497 Chr.17: 74918063-74918063 on Build GRCh38
rs924003689 Chr.17: 74918039-74918039 on Build GRCh38
rs139185240 Chr.17: 74918147-74918147 on Build GRCh38
rs376850581 Chr.17: 74917983-74917983 on Build GRCh38
rs1005613359 Chr.17: 74917935-74917935 on Build GRCh38
rs767766308 Chr.17: 74918035-74918035 on Build GRCh38
rs527247025 Chr.17: 74917979-74917979 on Build GRCh38
rs546608451 Chr.17: 74918112-74918112 on Build GRCh38
rs779827463 Chr.17: 74918077-74918077 on Build GRCh38
rs529630489 Chr.17: 74918009-74918009 on Build GRCh38
rs958836687 Chr.17: 74917992-74917992 on Build GRCh38
rs775570788 Chr.17: 74918055-74918055 on Build GRCh38
rs754803343 Chr.17: 74918085-74918085 on Build GRCh38
rs569986070 Chr.17: 74917998-74917998 on Build GRCh38
rs563126068 Chr.17: 74918015-74918015 on Build GRCh38
rs998184111 Chr.17: 74918020-74918020 on Build GRCh38
rs754374776 Chr.17: 74918066-74918066 on Build GRCh38
rs372068582 Chr.17: 74918070-74918070 on Build GRCh38
rs199648830 Chr.17: 74918024-74918024 on Build GRCh38
rs368597653 Chr.17: 74918118-74918118 on Build GRCh38
rs547358972 Chr.17: 74918080-74918080 on Build GRCh38
rs181973786 Chr.17: 74917962-74917962 on Build GRCh38
rs368231718 Chr.17: 74918060-74918060 on Build GRCh38
rs111415827 Chr.17: 74918096-74918096 on Build GRCh38
rs886053388 Chr.17: 74917978-74917978 on Build GRCh38
rs756229183 Chr.17: 74918022-74918022 on Build GRCh38

View Full Product Details