Assay Design



Gene Transcripts

Gene Symbol
IRX3
Entrez Gene ID
79191
Gene Name
iroquois homeobox 3
Gene Aliases
IRX-1, IRXB1
Chromosome Location
Chr.16: 54283300 - 54286772 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024336.2 NP_077312.2 3 - 4 1867 85
GenBank mRNA AY335943.1 - 3 - 4 1867 85
BC023667.2 - 3 - 4 1834 85
BC034328.1 - 2 - 3 547 85

View Full Product Details