Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020936.2 | NP_065987.1 | 10 - 11 | 1121 | 73 |
NR_135104.1 | 10 - 11 | 1121 | 73 | ||
NR_135105.1 | 10 - 11 | 1121 | 73 | ||
XM_017019710.1 | XP_016875199.1 | 10 - 11 | 1160 | 73 | |
XM_017019711.1 | XP_016875200.1 | 9 - 10 | 1215 | 73 | |
XM_017019712.1 | XP_016875201.1 | 10 - 11 | 1013 | 73 | |
XM_017019713.1 | XP_016875202.1 | 10 - 11 | 1136 | 73 | |
XM_017019714.1 | XP_016875203.1 | 11 - 12 | 1294 | 73 | |
XM_017019717.1 | XP_016875206.1 | 10 - 11 | 1058 | 73 | |
XM_017019718.1 | XP_016875207.1 | 8 - 9 | 1476 | 73 | |
XM_017019719.1 | XP_016875208.1 | 10 - 11 | 1013 | 73 | |
XM_017019720.1 | XP_016875209.1 | 8 - 9 | 2091 | 73 | |
GenBank mRNA | AB046815.1 | - | 5 - 6 | 2948 | 73 |
AK131442.1 | - | 7 - 8 | 1360 | 73 | |
AK301043.1 | - | 10 - 11 | 1084 | 73 | |
AK307384.1 | - | 10 - 11 | 1097 | 73 | |
AL833756.1 | - | 8 - 9 | 2158 | 73 | |
BC030020.2 | - | 10 - 11 | 1065 | 73 |