Assay Design



Gene Transcripts

Gene Symbol
MTHFD2L
Entrez Gene ID
441024
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like
Gene Aliases
-
Chromosome Location
Chr.4: 74123280 - 74303099 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017008231.1 XP_016863720.1 2 - 3 223 127
GenBank mRNA BC032771.1 - 1 - 2 96 127

View Full Product Details