Assay Design
Gene Transcripts
- Gene Symbol
- SLC25A42
- Entrez Gene ID
-
284439
- Gene Name
- solute carrier family 25 member 42
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 19063961 - 19113030 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178526.4 |
NP_848621.2 |
1 - 2 |
117 |
142 |
GenBank mRNA |
BC045598.1 |
- |
1 - 2 |
114 |
142 |
View Full Product Details