Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178498.3 | NP_848593.2 | 5 - 6 | 997 | 143 |
XM_006718155.3 | XP_006718218.1 | 5 - 6 | 900 | 143 | |
XM_006718156.3 | XP_006718219.1 | 5 - 6 | 1236 | 143 | |
XM_011519920.2 | XP_011518222.1 | 6 - 7 | 477 | 143 | |
XM_011519921.2 | XP_011518223.1 | 5 - 6 | 1237 | 143 | |
XM_017017244.1 | XP_016872733.1 | 6 - 7 | 923 | 143 | |
GenBank mRNA | BC041454.1 | - | 5 - 6 | 466 | 143 |
BC049207.1 | - | 5 - 6 | 969 | 143 |