Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004421.2 | NP_004412.2 | 14 - 15 | 1683 | 51 |
XM_005244731.3 | XP_005244788.1 | 14 - 15 | 1788 | 51 | |
GenBank mRNA | AF006011.1 | - | 14 - 15 | 1639 | 51 |
AK093189.1 | - | 7 - 8 | 923 | 51 | |
AK095867.1 | - | 7 - 8 | 901 | 51 | |
AK293286.1 | - | 14 - 15 | 1489 | 51 | |
AK300063.1 | - | 6 - 7 | 685 | 51 | |
BC017225.2 | - | 12 - 13 | 1408 | 51 | |
BC050454.1 | - | 8 - 9 | 1028 | 51 | |
U46461.1 | - | 14 - 15 | 1780 | 51 |