Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242339.1 | NP_001229268.1 | 3 - 4 | 306 | 78 |
NM_001323067.1 | NP_001309996.1 | 2 - 3 | 125 | 78 | |
NM_001323068.1 | NP_001309997.1 | 2 - 3 | 262 | 78 | |
NM_001323069.1 | NP_001309998.1 | 2 - 3 | 262 | 78 | |
NM_001323071.1 | NP_001310000.1 | 3 - 4 | 378 | 78 | |
NM_001323072.1 | NP_001310001.1 | 3 - 4 | 485 | 78 | |
NM_002627.4 | NP_002618.1 | 2 - 3 | 262 | 78 | |
XM_005252465.4 | XP_005252522.1 | 2 - 3 | 307 | 78 | |
XM_005252466.4 | XP_005252523.1 | 2 - 3 | 307 | 78 | |
XM_005252467.2 | XP_005252524.1 | 2 - 3 | 231 | 78 | |
XM_006717449.1 | XP_006717512.1 | 2 - 3 | 231 | 78 | |
GenBank mRNA | AK092597.1 | - | 3 - 4 | 306 | 78 |
AK291841.1 | - | 2 - 3 | 234 | 78 | |
AK308003.1 | - | 2 - 3 | 226 | 78 | |
BC002536.1 | - | 2 - 3 | 222 | 78 | |
BC029138.1 | - | 2 - 3 | 262 | 78 | |
D25328.1 | - | 2 - 3 | 219 | 78 |