Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282556.1 | NP_001269485.1 | 9 - 10 | 1447 | 85 |
NM_001282557.1 | NP_001269486.1 | 9 - 10 | 1378 | 85 | |
NM_001282558.1 | NP_001269487.1 | 9 - 10 | 1474 | 85 | |
NM_001282559.1 | NP_001269488.1 | 10 - 11 | 1263 | 85 | |
NM_007072.3 | NP_009003.1 | 9 - 10 | 1638 | 85 | |
XM_005247080.3 | XP_005247137.1 | 10 - 11 | 1508 | 85 | |
XM_011512367.2 | XP_011510669.1 | 9 - 10 | 1403 | 85 | |
XM_017005642.1 | XP_016861131.1 | 9 - 10 | 1438 | 85 | |
GenBank mRNA | AF126162.1 | - | 9 - 10 | 1378 | 85 |
AK000692.1 | - | 8 - 9 | 1373 | 85 | |
BC035971.1 | - | 9 - 10 | 1638 | 85 |