Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015590.3 | NP_056405.2 | 7 - 8 | 540 | 66 |
NM_182679.2 | NP_872620.1 | 7 - 8 | 614 | 66 | |
XM_005245287.3 | XP_005245344.1 | 6 - 7 | 536 | 66 | |
GenBank mRNA | AK000256.1 | - | 7 - 8 | 607 | 66 |
AK125763.1 | - | 4 - 5 | 2334 | 66 | |
AK293026.1 | - | 7 - 8 | 608 | 66 | |
BC032436.1 | - | 7 - 8 | 593 | 66 | |
BC040147.1 | - | 7 - 8 | 597 | 66 |