Assay Design



Gene Transcripts

Gene Symbol
HYAL1
Entrez Gene ID
3373
Gene Name
hyaluronoglucosaminidase 1
Gene Aliases
HYAL-1, LUCA1, MPS9, NAT6
Chromosome Location
Chr.3: 50299889 - 50312951 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153282.2 NP_695014.1 2 - 3 1057 72
GenBank mRNA AF502904.1 - 2 - 3 1029 72
CR541933.1 - 1 - 2 897 72
KU178143.1 - 1 - 2 897 72

View Full Product Details