Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144601.3 | NP_653202.1 | 3 - 4 | 610 | 74 |
NM_181553.2 | NP_853531.1 | 2 - 3 | 446 | 74 | |
NR_037613.1 | 3 - 4 | 830 | 74 | ||
NR_037619.1 | 2 - 3 | 379 | 74 | ||
XM_006721131.1 | XP_006721194.1 | 3 - 4 | 654 | 74 | |
GenBank mRNA | AF479813.1 | - | 3 - 4 | 584 | 74 |
AK056324.1 | - | 3 - 4 | 830 | 74 | |
AK075075.1 | - | 1 - 2 | 143 | 74 | |
BC023509.1 | - | 2 - 3 | 352 | 74 | |
BC023591.2 | - | 2 - 3 | 344 | 74 | |
BC036042.1 | - | 2 - 3 | 406 | 74 | |
BC040227.1 | - | 2 - 3 | 397 | 74 | |
BC047701.1 | - | 2 - 3 | 446 | 74 | |
BI457147.1 | - | 2 - 3 | 298 | 74 | |
CT001840.1 | - | 2 - 3 | 369 | 74 |