Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95845040-95845285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR2
Entrez Gene ID
8898
Gene Name
myotubularin related protein 2
Gene Aliases
CMT4B, CMT4B1
UniGene
Hs.181326
Gene Chromosome Location
Chr.11: 95832880-95924207 on Build GRCh38
Transcripts
NM_001243571.1 NM_016156.5 NM_201278.2 NM_201281.2 XM_005274374.2 XM_005274375.2 XM_006718934.2 XM_006718935.2 XM_006718936.3 XM_011543058.2 XM_011543059.2 XM_017018517.1 XM_017018518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6045675 c.1217T>C p.V406A Chr.11: 95845122-95845122 on Build GRCh38
933621 c.1052G>A p.R351H Chr.11: 95845071-95845071 on Build GRCh38
223492 c.1012A>G p.K338E Chr.11: 95845111-95845111 on Build GRCh38
3810725 c.1083C>A p.L361L Chr.11: 95845040-95845040 on Build GRCh38
933623 c.1016C>T p.T339M Chr.11: 95845107-95845107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773907921 Chr.11: 95845164-95845165 on Build GRCh38
rs113897932 Chr.11: 95845106-95845106 on Build GRCh38
rs780814285 Chr.11: 95845179-95845179 on Build GRCh38
rs779381469 Chr.11: 95845167-95845167 on Build GRCh38
rs780756016 Chr.11: 95845114-95845114 on Build GRCh38
rs777000344 Chr.11: 95845162-95845163 on Build GRCh38
rs1054904789 Chr.11: 95845226-95845226 on Build GRCh38
rs751057362 Chr.11: 95845174-95845174 on Build GRCh38
rs747853106 Chr.11: 95845184-95845184 on Build GRCh38
rs754483186 Chr.11: 95845176-95845176 on Build GRCh38
rs145714734 Chr.11: 95845149-95845149 on Build GRCh38
rs755844108 Chr.11: 95845195-95845195 on Build GRCh38
rs746601025 Chr.11: 95845100-95845100 on Build GRCh38
rs747904048 Chr.11: 95845115-95845115 on Build GRCh38
rs755349745 Chr.11: 95845224-95845224 on Build GRCh38
rs767174660 Chr.11: 95845186-95845186 on Build GRCh38
rs1037261406 Chr.11: 95845247-95845249 on Build GRCh38
rs762894011 Chr.11: 95845141-95845141 on Build GRCh38
rs761119018 Chr.11: 95845164-95845164 on Build GRCh38
rs377452665 Chr.11: 95845062-95845062 on Build GRCh38
rs121434402 Chr.11: 95845063-95845063 on Build GRCh38
rs777551816 Chr.11: 95845203-95845203 on Build GRCh38
rs896640684 Chr.11: 95845231-95845231 on Build GRCh38
rs148890896 Chr.11: 95845156-95845156 on Build GRCh38
rs773118782 Chr.11: 95845134-95845134 on Build GRCh38
rs757834677 Chr.11: 95845090-95845090 on Build GRCh38
rs769640482 Chr.11: 95845120-95845120 on Build GRCh38
rs972571750 Chr.11: 95845118-95845118 on Build GRCh38
rs779618687 Chr.11: 95845091-95845091 on Build GRCh38
rs751941205 Chr.11: 95845214-95845214 on Build GRCh38
rs754465336 Chr.11: 95845079-95845079 on Build GRCh38
rs925005819 Chr.11: 95845177-95845177 on Build GRCh38
rs1013772073 Chr.11: 95845233-95845233 on Build GRCh38
rs200313472 Chr.11: 95845103-95845103 on Build GRCh38
rs774525225 Chr.11: 95845155-95845155 on Build GRCh38
rs770852495 Chr.11: 95845152-95845152 on Build GRCh38
rs767757283 Chr.11: 95845163-95845163 on Build GRCh38
rs757812532 Chr.11: 95845166-95845166 on Build GRCh38
rs754328484 Chr.11: 95845165-95845165 on Build GRCh38

View Full Product Details