Assay Design
Gene Transcripts
- Gene Symbol
- CSHL1
- Entrez Gene ID
-
1444
- Gene Name
- chorionic somatomammotropin hormone like 1
- Gene Aliases
-
CS-5, CSHP1, CSL, GHB4, hCS-L
- Chromosome Location
-
Chr.17: 63909597 - 63911328 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_022581.2 |
NP_072103.1 |
2 - 3 |
323 |
153 |
View Full Product Details