Assay Design
Gene Transcripts
- Gene Symbol
- HYPK
- Entrez Gene ID
-
25764
- Gene Name
- huntingtin interacting protein K
- Gene Aliases
-
C15orf63, HSPC136
- Chromosome Location
-
Chr.15: 43800421 - 43802589 on Build GRCh38
- Gene Symbol
- SERF2-C15ORF63
- Entrez Gene ID
-
100529067
- Gene Name
- SERF2-C15orf63 readthrough
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 43791976 - 43802589 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037673.1 |
|
6 - 6 |
1376 |
141 |
View Full Product Details