Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002157.2 | NP_002148.1 | 1 - 1 | 381 | 68 |
GenBank mRNA | BC023518.2 | - | 1 - 1 | 55 | 68 |
CN264788.1 | - | 1 - 1 | 386 | 68 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202485.1 | NP_001189414.1 | 1 - 1 | 381 | 68 |