Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50956207-50956432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
UniGene
Hs.306291
Gene Chromosome Location
Chr.13: 50909678-50970462 on Build GRCh38
Transcripts
NM_001142279.2 NM_024570.3 XM_006719867.3 XM_011535229.1 XM_011535233.2 XM_017020747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1677671 c.838A>G p.T280A Chr.13: 50956373-50956373 on Build GRCh38
4967737 c.859G>T p.A287S Chr.13: 50956394-50956394 on Build GRCh38
1629142 c.860C>G p.A287G Chr.13: 50956395-50956395 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200802557 Chr.13: 50956365-50956365 on Build GRCh38
rs758605855 Chr.13: 50956389-50956389 on Build GRCh38
rs144408326 Chr.13: 50956394-50956394 on Build GRCh38
rs113527942 Chr.13: 50956325-50956325 on Build GRCh38
rs1039535362 Chr.13: 50956245-50956245 on Build GRCh38
rs775435037 Chr.13: 50956356-50956357 on Build GRCh38
rs746868812 Chr.13: 50956357-50956358 on Build GRCh38
rs773982703 Chr.13: 50956341-50956341 on Build GRCh38
rs750279427 Chr.13: 50956369-50956369 on Build GRCh38
rs906615816 Chr.13: 50956335-50956335 on Build GRCh38
rs942189407 Chr.13: 50956396-50956396 on Build GRCh38
rs766157957 Chr.13: 50956383-50956383 on Build GRCh38
rs1328360 Chr.13: 50956278-50956278 on Build GRCh38
rs559000645 Chr.13: 50956360-50956360 on Build GRCh38
rs771820918 Chr.13: 50956342-50956343 on Build GRCh38
rs769157680 Chr.13: 50956260-50956260 on Build GRCh38
rs151005517 Chr.13: 50956283-50956283 on Build GRCh38
rs760409807 Chr.13: 50956378-50956378 on Build GRCh38
rs761906095 Chr.13: 50956359-50956359 on Build GRCh38
rs775040772 Chr.13: 50956315-50956315 on Build GRCh38
rs1009541501 Chr.13: 50956286-50956286 on Build GRCh38
rs752930177 Chr.13: 50956388-50956388 on Build GRCh38
rs950747194 Chr.13: 50956363-50956363 on Build GRCh38
rs936576461 Chr.13: 50956274-50956274 on Build GRCh38
rs998580144 Chr.13: 50956299-50956299 on Build GRCh38
rs921212689 Chr.13: 50956246-50956246 on Build GRCh38
rs957421838 Chr.13: 50956320-50956320 on Build GRCh38
rs768290667 Chr.13: 50956328-50956328 on Build GRCh38
rs746198570 Chr.13: 50956307-50956307 on Build GRCh38
rs367543519 Chr.13: 50956312-50956312 on Build GRCh38
rs900791137 Chr.13: 50956276-50956276 on Build GRCh38

View Full Product Details