Assay Design
Gene Transcripts
- Gene Symbol
- C17orf82
- Entrez Gene ID
-
388407
- Gene Name
- chromosome 17 open reading frame 82
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 61411751 - 61413280 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_203425.2 |
NP_982249.2 |
1 - 1 |
1454 |
129 |
GenBank mRNA |
BC046200.1 |
- |
1 - 1 |
1454 |
129 |
View Full Product Details