Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005666.3 | NP_005657.1 | 3 - 4 | 542 | 114 |
XM_005245113.3 | XP_005245170.1 | 2 - 3 | 361 | 114 | |
XM_011509459.2 | XP_011507761.1 | 3 - 4 | 576 | 114 | |
GenBank mRNA | BC022283.1 | - | 3 - 4 | 531 | 114 |
X64877.1 | - | 3 - 4 | 506 | 114 |