Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47215040-47215309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB2
Entrez Gene ID
1643
Gene Name
damage specific DNA binding protein 2
Gene Aliases
DDBB, UV-DDB2, XPE
UniGene
Hs.700338
Gene Chromosome Location
Chr.11: 47214942-47239218 on Build GRCh38
Transcripts
NM_000107.2 NM_001300734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7130959 c.111C>G p.L37L Chr.11: 47215247-47215247 on Build GRCh38
5895260 c.53_54insC p.R20fs*26 Chr.11: 47215189-47215190 on Build GRCh38
1981286 c.43A>C p.I15L Chr.11: 47215179-47215179 on Build GRCh38
6581410 c.126C>G p.S42S Chr.11: 47215262-47215262 on Build GRCh38
6641244 c.2T>A p.M1K Chr.11: 47215138-47215138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780921271 Chr.11: 47215108-47215108 on Build GRCh38
rs750851705 Chr.11: 47215190-47215190 on Build GRCh38
rs530733334 Chr.11: 47215240-47215240 on Build GRCh38
rs943544509 Chr.11: 47215194-47215194 on Build GRCh38
rs367717420 Chr.11: 47215116-47215116 on Build GRCh38
rs1051950278 Chr.11: 47215154-47215154 on Build GRCh38
rs561846496 Chr.11: 47215123-47215124 on Build GRCh38
rs201317629 Chr.11: 47215101-47215101 on Build GRCh38
rs148175447 Chr.11: 47215215-47215215 on Build GRCh38
rs777698820 Chr.11: 47215217-47215217 on Build GRCh38
rs768981688 Chr.11: 47215072-47215072 on Build GRCh38
rs764220947 Chr.11: 47215108-47215109 on Build GRCh38
rs371281878 Chr.11: 47215127-47215127 on Build GRCh38
rs375840702 Chr.11: 47215106-47215106 on Build GRCh38
rs369110013 Chr.11: 47215156-47215156 on Build GRCh38
rs772542105 Chr.11: 47215257-47215257 on Build GRCh38
rs201229167 Chr.11: 47215188-47215188 on Build GRCh38
rs956219237 Chr.11: 47215205-47215205 on Build GRCh38
rs180861241 Chr.11: 47215092-47215092 on Build GRCh38
rs954238770 Chr.11: 47215126-47215126 on Build GRCh38
rs199965459 Chr.11: 47215268-47215268 on Build GRCh38
rs749034260 Chr.11: 47215219-47215219 on Build GRCh38
rs754254385 Chr.11: 47215165-47215165 on Build GRCh38
rs775814662 Chr.11: 47215239-47215239 on Build GRCh38
rs777159854 Chr.11: 47215081-47215081 on Build GRCh38
rs765788313 Chr.11: 47215282-47215282 on Build GRCh38
rs760606007 Chr.11: 47215143-47215143 on Build GRCh38
rs754405766 Chr.11: 47215199-47215199 on Build GRCh38
rs748147583 Chr.11: 47215112-47215112 on Build GRCh38
rs373622283 Chr.11: 47215195-47215195 on Build GRCh38
rs762435826 Chr.11: 47215167-47215167 on Build GRCh38
rs779621308 Chr.11: 47215193-47215193 on Build GRCh38
rs758089001 Chr.11: 47215100-47215100 on Build GRCh38
rs4647708 Chr.11: 47215090-47215090 on Build GRCh38
rs765557363 Chr.11: 47215189-47215189 on Build GRCh38
rs772481534 Chr.11: 47215123-47215123 on Build GRCh38
rs376193077 Chr.11: 47215128-47215128 on Build GRCh38
rs889406597 Chr.11: 47215177-47215177 on Build GRCh38
rs770855579 Chr.11: 47215233-47215233 on Build GRCh38
rs776815495 Chr.11: 47215258-47215258 on Build GRCh38
rs368073045 Chr.11: 47215248-47215248 on Build GRCh38
rs568689968 Chr.11: 47215191-47215191 on Build GRCh38
rs757228612 Chr.11: 47215124-47215124 on Build GRCh38
rs759835280 Chr.11: 47215122-47215122 on Build GRCh38
rs773718543 Chr.11: 47215283-47215283 on Build GRCh38
rs780776638 Chr.11: 47215213-47215213 on Build GRCh38
rs769425973 Chr.11: 47215115-47215115 on Build GRCh38

View Full Product Details