Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67230077-67230332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
UniGene
Hs.95231
Gene Chromosome Location
Chr.16: 67229389-67247522 on Build GRCh38
Transcripts
NM_001318202.1 NM_013241.2 XM_006721180.1 XM_011523043.2 XM_011523044.1 XM_011523045.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3370501 c.3037C>T p.R1013C Chr.16: 67230328-67230328 on Build GRCh38
6571421 c.3057G>C p.E1019D Chr.16: 67230223-67230223 on Build GRCh38
6663964 c.3178A>G p.R1060G Chr.16: 67230102-67230102 on Build GRCh38
5980998 c.3100G>T p.V1034F Chr.16: 67230180-67230180 on Build GRCh38
972357 c.3063C>A p.F1021L Chr.16: 67230217-67230217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764063407 Chr.16: 67230270-67230270 on Build GRCh38
rs751257468 Chr.16: 67230228-67230228 on Build GRCh38
rs373759873 Chr.16: 67230280-67230280 on Build GRCh38
rs376653596 Chr.16: 67230122-67230122 on Build GRCh38
rs762977687 Chr.16: 67230237-67230238 on Build GRCh38
rs775428830 Chr.16: 67230262-67230262 on Build GRCh38
rs780524217 Chr.16: 67230309-67230309 on Build GRCh38
rs557236288 Chr.16: 67230186-67230186 on Build GRCh38
rs754951818 Chr.16: 67230108-67230108 on Build GRCh38
rs201129113 Chr.16: 67230217-67230217 on Build GRCh38
rs368888802 Chr.16: 67230142-67230142 on Build GRCh38
rs768810239 Chr.16: 67230250-67230250 on Build GRCh38
rs373077110 Chr.16: 67230224-67230224 on Build GRCh38
rs146514223 Chr.16: 67230180-67230180 on Build GRCh38
rs767751100 Chr.16: 67230197-67230197 on Build GRCh38
rs776135636 Chr.16: 67230162-67230162 on Build GRCh38
rs759335200 Chr.16: 67230258-67230258 on Build GRCh38
rs752564255 Chr.16: 67230206-67230206 on Build GRCh38
rs781372321 Chr.16: 67230156-67230156 on Build GRCh38
rs373710737 Chr.16: 67230150-67230150 on Build GRCh38
rs757844941 Chr.16: 67230125-67230125 on Build GRCh38
rs759317021 Chr.16: 67230200-67230200 on Build GRCh38
rs780976238 Chr.16: 67230232-67230232 on Build GRCh38
rs769846134 Chr.16: 67230200-67230201 on Build GRCh38
rs886507546 Chr.16: 67230168-67230168 on Build GRCh38
rs561555976 Chr.16: 67230304-67230304 on Build GRCh38
rs776409748 Chr.16: 67230261-67230261 on Build GRCh38
rs780716486 Chr.16: 67230248-67230248 on Build GRCh38
rs746584932 Chr.16: 67230149-67230149 on Build GRCh38
rs867548759 Chr.16: 67230212-67230212 on Build GRCh38
rs749705095 Chr.16: 67230249-67230249 on Build GRCh38
rs988685147 Chr.16: 67230292-67230292 on Build GRCh38
rs756907127 Chr.16: 67230229-67230229 on Build GRCh38
rs990689517 Chr.16: 67230215-67230215 on Build GRCh38
rs983111425 Chr.16: 67230153-67230153 on Build GRCh38
rs746301227 Chr.16: 67230234-67230234 on Build GRCh38
rs546547630 Chr.16: 67230160-67230160 on Build GRCh38
rs1049123335 Chr.16: 67230161-67230161 on Build GRCh38
rs142647972 Chr.16: 67230123-67230123 on Build GRCh38
rs768557197 Chr.16: 67230178-67230178 on Build GRCh38
rs376362919 Chr.16: 67230307-67230307 on Build GRCh38
rs916425990 Chr.16: 67230235-67230235 on Build GRCh38
rs996408352 Chr.16: 67230225-67230225 on Build GRCh38
rs528954834 Chr.16: 67230310-67230310 on Build GRCh38
rs773422266 Chr.16: 67230257-67230257 on Build GRCh38
rs762761528 Chr.16: 67230269-67230269 on Build GRCh38
rs773360285 Chr.16: 67230236-67230238 on Build GRCh38
rs1040516891 Chr.16: 67230195-67230195 on Build GRCh38
rs772244488 Chr.16: 67230253-67230253 on Build GRCh38
rs765229597 Chr.16: 67230203-67230203 on Build GRCh38
rs770216033 Chr.16: 67230244-67230244 on Build GRCh38
rs753800789 Chr.16: 67230099-67230099 on Build GRCh38
rs767282545 Chr.16: 67230293-67230293 on Build GRCh38
rs949948364 Chr.16: 67230139-67230139 on Build GRCh38
rs908962749 Chr.16: 67230167-67230167 on Build GRCh38
rs955879919 Chr.16: 67230112-67230112 on Build GRCh38
rs188445865 Chr.16: 67230251-67230251 on Build GRCh38
rs762098881 Chr.16: 67230192-67230193 on Build GRCh38
rs751086498 Chr.16: 67230271-67230271 on Build GRCh38
rs577832197 Chr.16: 67230159-67230159 on Build GRCh38
rs935338314 Chr.16: 67230299-67230299 on Build GRCh38
rs763128197 Chr.16: 67230174-67230174 on Build GRCh38

View Full Product Details