Assay Design



Gene Transcripts

Gene Symbol
CLIC4
Entrez Gene ID
25932
Gene Name
chloride intracellular channel 4
Gene Aliases
CLIC4L, H1, MTCLIC, huH1, p64H1
Chromosome Location
Chr.1: 24745269 - 24844324 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013943.2 NP_039234.1 6 - 6 2842 140
GenBank mRNA AL080061.1 - 1 - 1 313 140
AL117424.1 - 6 - 6 2754 140

View Full Product Details