Assay Design



Gene Transcripts

Gene Symbol
SOCS5
Entrez Gene ID
9655
Gene Name
suppressor of cytokine signaling 5
Gene Aliases
CIS6, CISH6, Cish5, SOCS-5
Chromosome Location
Chr.2: 46698960 - 46762788 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014011.4 NP_054730.1 2 - 2 4352 95
NM_144949.2 NP_659198.1 2 - 2 4440 95
GenBank mRNA AB014571.1 - 2 - 2 4360 95

View Full Product Details