Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152288.2 | NP_689501.1 | 2 - 2 | 2057 | 153 |
GenBank mRNA | BC006126.1 | - | 2 - 2 | 1999 | 153 |
BC015555.1 | - | 2 - 2 | 2045 | 153 | |
BC022786.1 | - | 2 - 2 | 1999 | 153 |